missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo Iii, Honeywell Fluka™

Product Code. 15671020 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15671020

Brand: Honeywell-Fluka 110905G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

for spectrophotometric det. of Th, Zr, U, Cd, Zn, Ca

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Linear Formula (HO)2C10H2(SO3H)2(N=NC6H4AsO3H2)2
UN Number 3465
Beilstein 5717957
Formula Weight 776.37g/mol
Packaging Glass Bottle
Quantity 5 g
Chemical Name or Material Arsenazo Iii
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.