Learn More
Invitrogen™ ARSA Recombinant Mouse Monoclonal Antibody (K29073_19C6)
Description
Immunogen sequence: RSGRGPFGDS LMELDAAVGT LMTAIGDLGL LEETLVIFTA DNGPETMRMS RGGCSGLLRC GKGTTYEGGV REPALAFWPG HIAPGVTHEL ASSLDLLPTL AALAGAPLPN VTLDGFDLSP LLLGTGKSPR QSLFFYPSYP DEVRGVFAVR TGKYKAHFFT QGSAHSDTTA DPACHASSSL TAHEPPLLYD LSKDPGENYN LLGGVAGATP EVLQALKQLQ LLKAQLDAAV TFGPSQVARG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 39%, Pig - 91%, Cynomolgus monkey - 97%.
Specifications
Specifications
| Antigen | ARSA |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K29073_19C6 |
| Concentration | 0.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | ARSA |
| Gene Accession No. | P15289 |
| Gene Alias | ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.