missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARSA Recombinant Mouse Monoclonal Antibody (K29073_19C6)

Product Code. 30824808
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824808 100 μL
30830342 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824808 Supplier Invitrogen™ Supplier No. MA564493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: RSGRGPFGDS LMELDAAVGT LMTAIGDLGL LEETLVIFTA DNGPETMRMS RGGCSGLLRC GKGTTYEGGV REPALAFWPG HIAPGVTHEL ASSLDLLPTL AALAGAPLPN VTLDGFDLSP LLLGTGKSPR QSLFFYPSYP DEVRGVFAVR TGKYKAHFFT QGSAHSDTTA DPACHASSSL TAHEPPLLYD LSKDPGENYN LLGGVAGATP EVLQALKQLQ LLKAQLDAAV TFGPSQVARG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 39%, Pig - 91%, Cynomolgus monkey - 97%.

The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARSA
Applications Western Blot
Classification Recombinant Monoclonal
Clone K29073_19C6
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ARSA
Gene Accession No. P15289
Gene Alias ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40
Gene Symbols ARSA
Host Species Mouse
Immunogen Recombinant human ARSA (aa 241-480).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 410
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.