Learn More
Invitrogen™ ARSA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595910
Description
Immunogen sequence: LMADAQRQDR PFFLYYASHH THYPQFSGQS FAERSGRGPF GDSLMELDAA VGTLMTAIGD LGLLEETLVI FTADNGPETM RMSRGGCSGL LRCGKGTTYE GGVREPALAF WPGHIAPGVT HELASSLDLL PTLAALAGAP LPNVTLDGFD LSPLLLGTGK SPRQSLFFYP SYPDEVRGVF AVRTGKYKAH FFTQGSAHSD TTADPACHAS SSLTAHEPPL LYDLSKDPGE NYNLLGGVAG ATPEVLQALK QLQLLKAQLD AAVTFGPSQV ARGEDPALQI CCHPGCTPRP ACCHCPDPHA; Positive Samples: Jurkat, HeLa, HepG2, A-549, Mouse testis, Mouse liver, Mouse brain; Cellular Location: Lysosome.
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications
| ARSA | |
| Polyclonal | |
| Unconjugated | |
| ARSA | |
| ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11883, 410 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15289, P50428 | |
| ARSA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-509 of human ARSA (NP_0004783). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.