missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARS2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-42501
This item is not returnable.
View return policy
Description
ARS2 Polyclonal antibody specifically detects ARS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA
Specifications
| ARS2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 51593 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Peptide ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 μg/mL, Peptide ELISA Detection limit 1:128000 | |
| ARS2serrate, arsenate resistance protein 2, arsenate resistance protein ARS2, arsenite resistance protein, Arsenite-resistance protein 2, Asr2, MGC126427, serrate RNA effector molecule homolog, serrate RNA effector molecule homolog (Arabidopsis) | |
| Peptide with sequence C-DPRAIVEYRDLDAPD corresponding to C-Terminus according to NP_001122324.1, NP_001122325.1, NP_001122326.1. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction