missing translation for 'onlineSavingsMsg'
Learn More

ARS2 Antibody, Novus Biologicals™

Product Code. 18372969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372969 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372969 Supplier Novus Biologicals Supplier No. H00051593B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ARS2 Polyclonal antibody specifically detects ARS2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARS2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10 to 1:500
Formulation PBS (pH 7.4)
Gene Alias ARS2serrate, arsenate resistance protein 2, arsenate resistance protein ARS2, arsenite resistance protein, Arsenite-resistance protein 2, Asr2, MGC126427, serrate RNA effector molecule homolog, serrate RNA effector molecule homolog (Arabidopsis)
Host Species Mouse
Immunogen ARS2 (NP_877952, 1 a.a. - 796 a.a.) full-length human protein. MRRDWDEHSSDPYHSGYEMPYAGGGGGPTYGPPQPWGHPDVHIMQHHVLPIQARLGSIAEIDLGVPPPVMKTFKEFLLSLDDSVDETEAVKRYNDYKLDFRRQQMQDFFLAHKDEEWFRSKYHPDEVGKRRQEARGALQNRLRVFLSLMETGWFDNLLLDIDKADAIVKMLDAAVIKMEGGTENDLRILEQEEEEEQAGKPGEPSKKEEGRAGAGLGDGERKTNDKDEKKEDGKQAENDSSNDDKTKKSEGDGDKEEKKEDSEKEAKKSSKKRNRKHSGDDSFDEGSVSESESESESGQAEEEKEEAEALKEKEKPKEEEWEKPKDAAGLECKPRPLHKTCSLFMRNIAPNISRAEIISLCKRYPGFMRVALSEPQPERRFFRRGWVTFDRSVNIKEICWNLQNIRLRECELSPGVNRDLTRRVRNINGITQHKQIVRNDIKLAAKLIHTLDDRTQLWASEPGTPPLPTSLPSQNPILKNITDYLIEEVSAEEEELLGSSGGAPPEEPPKEGNPAEINVERDEKLIKVLDKLLLYLRIVHSLDYYNTCEYPNEDEMPNRCGIIHVRGPMPPNRISHGEVLEWQKTFEEKLTPLLSVRESLSEEEAQKMGRKDPEQEVEKFVTSNTQELGKDKWLCPLSGKKFKGPEFVRKHIFNKHAEKIEEVKKEVAFFNNFLTDAKRPALPEIKPAQPPGPAQILPPGLTPGLPYPHQTPQGLMPYGQPRPPILGYGAGAVRPAVPTGGPPYPHAPYGAGRGNYDAFRGQGGYPGKPRNRMVRGDPRAIVEYRDLDAPDDVDFF
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51593
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.