missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARS-853, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor with an IC50 of 2.5 μM. ARS-853 inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation.
Specifications
Specifications
| Chemical Name or Material | ARS-853 |
| CAS | 1629268-00-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H29ClN4O3 |
| Quantity | 1 mg |
| Solubility Information | DMSO : 33.33 mg/mL (76.99 mM; Need ultrasonic) |
| SMILES | ClC1=C(C2(C)CC2)C=C(NCC(N3CCN(C4CN(C(C=C)=O)C4)CC3)=O)C(O)=C1 |
| Molecular Weight (g/mol) | 432.94 |
| Formula Weight | 432.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction