missing translation for 'onlineSavingsMsg'
Learn More

ARS-853, MedChemExpress

Product Code. 16493797
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493797 50 mg 50mg
16443676 25 mg 25mg
16440318 5 mg 5mg
16431849 1 mg 1mg
16407858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493797 Supplier Medchem Express Supplier No. HY19706
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor with an IC50 of 2.5 μM. ARS-853 inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation.

ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor with an IC50 of 2.5 μM. ARS-853 inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation.

Specifications

Chemical Name or Material ARS-853
CAS 1629268-00-3
Color White
Purity Grade Notes Research
Molecular Formula C22H29ClN4O3
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (76.99 mM; Need ultrasonic)
SMILES ClC1=C(C2(C)CC2)C=C(NCC(N3CCN(C4CN(C(C=C)=O)C4)CC3)=O)C(O)=C1
Molecular Weight (g/mol) 432.94
Formula Weight 432.94
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.