missing translation for 'onlineSavingsMsg'
Learn More

ARS-1620, MedChemExpress

Product Code. 16485437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485437

Brand: Medchem Express HYU00418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ARS-1620 is an atropisomeric selective KRASG12C inhibitor with desirable pharmacokinetics.

ARS-1620 is an atropisomeric selective KRASG12C inhibitor with desirable pharmacokinetics.

Specifications

Chemical Name or Material ARS-1620
CAS 1698055-85-4
Color White
Purity Grade Notes Research
Molecular Formula C21H17ClF2N4O2
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (290.13 mM; Need ultrasonic) ∣H2O : 11 mg/mL (25.53 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES C=CC(N1CCN(C2=C3C=C(Cl)[C@]([C@]4=C(O)C=CC=C4F)=C(F)C3=NC=N2)CC1)=O.[S]
Molecular Weight (g/mol) 430.84
Formula Weight 430.84
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.