missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARRY 520 trifluoroacetate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Beschreibung
Potent kinesin spindle protein (KSP) inhibitor (IC50 = 6 nM); selective for KSP over >250 other receptors and kinases at a concentration of 10 μM. Displays robust antitumor activity in bortezomib-resistant xenografts either alone or in combination with bortezomib. Induces degradation of Mcl-1; exhibits comparable cytotoxic activity to taxol (Cat. No. 1097) in epithelial ovarian cancer cells. Active in vivo.
Spezifikation
Spezifikation
CAS | 885060-09-3 |
Molecular Formula | C20H22F2N4O2S |
Quantity | 10 mg |
Synonym | arry-520, filanesib, unii-8a49oso368, arry 520 trifluoroacetate, 2s-2-3-aminopropyl-5-2,5-difluorophenyl-n-methoxy-n-methyl-2-phenyl-1,3,4-thiadiazole-3 2h-carboxamide, arry 520, filanesib inn, filanesib usan:inn, arry520, arry-520; filanesib |
InChI Key | LLXISKGBWFTGEI-FQEVSTJZSA-N |
SMILES | CN(C(=O)N1C(SC(=N1)C2=C(C=CC(=C2)F)F)(CCCN)C3=CC=CC=C3)OC |
IUPAC Name | (2S)-2-(3-aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3-carboxamide |
Molecular Weight (g/mol) | 420.479 |
PubChem CID | 44224257 |
Formula Weight | 420.48 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'