Learn More
Abeba™ Arrow 38210 Shoes
Product Code. 15661158
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15661158 44 1 pair
15681148 36 1 pair
15691148 37 1 pair
15601158 38 1 pair
15611158 39 1 pair
15621158 40 1 pair
15631158 41 1 pair
15641158 42 1 pair
15651158 43 1 pair
15671158 45 1 pair
15681158 46 1 pair
15691158 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15661158 Supplier Abeba™ Supplier No. 38210/44
  • ONLINE EXCLUSIVE PRICE
    £77.65
    £51.76 / 1 pairs
    Save £25.89
    33% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather
  • Black
  • Slip resistance PU/PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRA
Color Black
Liner Material Microfiber
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Polyurethane
Style 38210