missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arrestin C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11100-2-AP-20UL
This item is not returnable.
View return policy
Description
Arrestin-C, also known as retinal cone Arrestin-3, X-Arrestin or cArr, is a member of the Arrestin family of proteins. It is predomitly found in the retina and pineal gland and localizes to the inner and outer segments of red-, green- and blue-cone photoreceptors and the inner plexiform regions. Two Arrestin-C isoforms exist due to alternative splicing. Isoform 1 is the mature full length protein and isoform 2 is truncated, ending with an arginine for amino acid residue 359. Arrestin-C expression is stimulated by retinoic acid. It may play a role in retina-specific signal transduction and bind to photoactivated-phosphorylated red/green opsins. In addition, Arrestin-C forms homodimers and oligomers with beta-Arrestins and may regulate beta-Arrestin mediated signaling.Specifications
| Arrestin C | |
| Polyclonal | |
| Unconjugated | |
| ARR3 | |
| ARR3, Arrestin C, ARRX, C arrestin, CAR, cArr, Cone arrestin, Retinal cone arrestin 3, X arrestin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170735, 407 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36575, Q9EQP6 | |
| ARR3 | |
| Arrestin C Fusion Protein Ag1580 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction