missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arrestin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Arrestin 3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Arrestin 3 Polyclonal specifically detects Arrestin 3 in Human samples. It is validated for Western Blot.Specifications
| Arrestin 3 | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision | |
| arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin | |
| ARR3 | |
| IgG | |
| 43 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 407 | |
| Synthetic peptides corresponding to ARR3 (arrestin 3, retinal (X-arrestin)) The peptide sequence was selected from the middle region of ARR3. Peptide sequence EDAGKPCGIDFEVKSFCAENPEETVSKRDYVRLVVRKVQFAPPEAGPGPS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title