missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARRDC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538852
This item is not returnable.
View return policy
Description
ARRDC1, ARRDC2 (which exists as multiple alternatively spliced isoforms), ARRDC4 and ARRDC5 are arrestin domain-containing proteins that are encoded by genes which map to human chromosomes 9, 15 and 19. Chromosome 9, on which the ARRDC1 gene is localized, contains 145 million base pairs and comprises 4% of the human genome, encoding nearly 900 genes. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and Familial dysautonomia, are both associated with chromosome 9.
Specifications
| ARRDC4 | |
| Polyclonal | |
| Unconjugated | |
| ARRDC4 | |
| Arrestin domain-containing protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 293019, 66412, 91947 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NCT1, A0A0B4J1F4, Q7TP90 | |
| ARRDC4 | |
| A synthetic peptide derived from the internal region of human ARRD4 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction