missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARRDC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536518
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 45 kDa. Purity is >95% by SDS-PAGE.
ARRDC1, ARRDC2 (which exists as multiple alternatively spliced isoforms), ARRDC4 and ARRDC5 are arrestin domain-containing proteins that are encoded by genes which map to human chromosomes 9, 15 and 19. Chromosome 9, on which the ARRDC1 gene is localized, contains 145 million base pairs and comprises 4% of the human genome, encoding nearly 900 genes. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and Familial dysautonomia, are both associated with chromosome 9.
Specifications
| ARRDC4 | |
| Polyclonal | |
| Unconjugated | |
| ARRDC4 | |
| 2410003C09Rik; Ab1-209; ARRDC4; arrestin domain containing 4; arrestin domain-containing protein 4; AV216361; liver regeneration-related protein LRRG041/LRRGT00117; LRRG00117 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91947 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NCT1 | |
| ARRDC4 | |
| Synthetic peptide corresponding to amino acids 130-180 of human Arrdc4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction