missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARRDC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | ARRDC3 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Specifications
| ARRDC3 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96B67 | |
| 57561 | |
| ARRDC3 Fusion Protein Ag18717 | |
| Primary | |
| -20°C | |
| ARRDC3 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ARRDC3, arrestin domain containing 3, KIAA1376, TLIMP | |
| ARRDC3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title