Description
- The Array Card RNaseP Verification Kit is used to verify your real-time PCR instrument with Array Card Block.
- Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.
- The Array Card RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
- The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well.
- To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
Specifications
Specifications
| Calibrated Dye | Proprietary Mix |
| For Use With (Equipment) | ViiA™ 7 Dx System, QuantStudio™ Dx |
| Product Type | Array Card RNAseP Verification Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Quantity | 1 array |
| Calibration Type | Instrument Verification Plate |
| PCR Method | qPCR |