missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The Array Card Background Buffer Kit is used to maintain your real-time PCR instrument with array card block.
- Background calibration should be run at least every six months.
- Please see your instrument user guide for full instructions on use.
Specifications
Specifications
| Content And Storage | Store Kit at or below -20°C. |
| For Use With (Equipment) | ViiA™ 7 Dx System, QuantStudio™ Dx |
| Product Type | Array Card Background Buffer Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Quantity | 6 cards kit |
| Calibration Type | Spectral Calibration |
| GC-Rich PCR Performance | Low |
| PCR Method | qPCR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?