missing translation for 'onlineSavingsMsg'
Learn More

ARQ 531, MedChemExpress

Product Code. 16495446
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495446 100 mg 100mg
16457317 5 mg 5mg
16443156 25 mg 25mg
16404608 10 mg 10mg
16400907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495446 Supplier Medchem Express Supplier No. HY112215
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ARQ 531 (MK-1026) is a reversible non-covalent and orally active inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively.

ARQ 531 (MK-1026) is a reversible non-covalent and orally active inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively.

Specifications

Chemical Name or Material ARQ 531
CAS 2095393-15-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H23ClN4O4
Quantity 100 mg
Synonym MK-1026
Solubility Information DMSO : ≥ 50 mg/mL (104.40 mM)
SMILES O=C(C1=C(C=C(OC2=CC=CC=C2)C=C1)Cl)C3=CNC4=NC=NC(N[C@@H]5CC[C@@H](CO)OC5)=C34
Molecular Weight (g/mol) 478.93
Formula Weight 478.93
Percent Purity 98.52%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.