missing translation for 'onlineSavingsMsg'
Learn More

ARQ 531, MedChemExpress

Product Code. 16443156
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443156

Brand: Medchem Express HY112215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ARQ 531 (MK-1026) is a reversible non-covalent and orally active inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively.

ARQ 531 (MK-1026) is a reversible non-covalent and orally active inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively.

Specifications

Chemical Name or Material ARQ 531
CAS 2095393-15-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H23ClN4O4
Quantity 25 mg
Synonym MK-1026
Solubility Information DMSO : ≥ 50 mg/mL (104.40 mM)
SMILES O=C(C1=C(C=C(OC2=CC=CC=C2)C=C1)Cl)C3=CNC4=NC=NC(N[C@@H]5CC[C@@H](CO)OC5)=C34
Molecular Weight (g/mol) 478.93
Formula Weight 478.93
Percent Purity 98.52%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.