missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPP-21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ARPP-21 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ARPP-21 Polyclonal specifically detects ARPP-21 in Mouse samples. It is validated for Western Blot.Specifications
| ARPP-21 | |
| Polyclonal | |
| Rabbit | |
| NP_001171090 | |
| 10777 | |
| Synthetic peptide towards ARPP-21. Peptide sequence SAPENGILKSESLDEEEKLELQRRLAAQNQERRKSKSGAGKGKLTRSLAV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 21 kD, cAMP-regulated phosphoprotein, 21kDa, R3H domain containing 3 | |
| ARPP21 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title