missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARPM1 Polyclonal specifically detects ARPM1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ARPM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | actin related protein M1, actin-related protein M1, MGC15664 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of mouse ARPM1 (NP_083966.1). Peptide sequence DVAKLAPANTAVQVIAPPERKISVWMGGSILASLSAFQDMWITAAEFEEV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?