missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARPC5 Recombinant Rabbit Monoclonal Antibody (K01_1L59)
GREENER_CHOICE

Product Code. 30827721
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827721 100 μL
30826694 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827721 Supplier Invitrogen™ Supplier No. MA563231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: SKNTVSSARF RKVDVDEYD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%, Cynomolgus monkey - 100%.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1L59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ARPC5
Gene Accession No. O15511, Q4KLF8, Q9CPW4
Gene Alias 5830443F10Rik; actin related protein 2/3 complex subunit 5; actin related protein 2/3 complex subunit 5, 16kDa; actin related protein 2/3 complex, subunit 5; actin related protein 2/3 complex, subunit 5, 16kDa; actin-related protein 2/3 complex subunit 5; actin-related protein 2/3 complex subunit 5-like protein; ARC16; Arp2/3 complex 16 kDa subunit; Arp2/3 protein complex subunit p16; Arpc5; dJ127C7.3; p16-ARC
Gene Symbols ARPC5
Host Species Rabbit
Immunogen A synthetic peptide of human p16 ARC (aa 2-20).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10092, 360854, 67771
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.