missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARPC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102886
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARPC5.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
Specifications
| ARPC5 | |
| Polyclonal | |
| Unconjugated | |
| ARPC5 | |
| 5830443F10Rik; actin related protein 2/3 complex subunit 5; actin related protein 2/3 complex subunit 5, 16kDa; actin related protein 2/3 complex, subunit 5; actin related protein 2/3 complex, subunit 5, 16kDa; actin-related protein 2/3 complex subunit 5; actin-related protein 2/3 complex subunit 5-like protein; ARC16; Arp2/3 complex 16 kDa subunit; Arp2/3 protein complex subunit p16; Arpc5; dJ127C7.3; p16-ARC | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10092 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15511 | |
| ARPC5 | |
| A synthesized peptide derived from human ARPC5(Accession O15511), corresponding to amino acid residues R10-K60. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction