missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550552
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ARPC4 protein.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.
Specifications
| ARPC4 | |
| Polyclonal | |
| Unconjugated | |
| ARPC4 | |
| 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10093, 68089 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P59998, P59999 | |
| ARPC4 | |
| Synthetic peptide corresponding to a region derived from internal residues of human actin related protein 2/3 complex, subunit 4, 20kDa. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction