missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ARPC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29634
This item is not returnable.
View return policy
Description
ARPC4 Polyclonal antibody specifically detects ARPC4 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ARPC4 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| actin related protein 2/3 complex, subunit 4, 20kDa, actin-related protein 2/3 complex subunit 4, arp2/3 protein complex subunit p20, MGC13544, p20-ARC, subunit 4 (20 kD) | |
| Between 1 and 50 | |
| 100 μg | |
| Protein Kinase | |
| 10093 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction