missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC2 Polyclonal Antibody

ARPC2 Polyclonal Antibody
Brand: Invitrogen PA588041
This item is not returnable.
View return policy
Description
ARPC2 Polyclonal Antibody for Western Blot
Specifications
| ARPC2 | |
| Polyclonal | |
| Unconjugated | |
| ARPC2 | |
| 2210023N03Rik; 34kDa; actin related protein 2/3 complex subunit 2; actin related protein 2/3 complex subunit 2, 34kDa; actin related protein 2/3 complex, subunit 2; actin related protein 2/3 complex, subunit 2, 34kDa; actin-related protein 2/3 complex subunit 2; actin-related protein 2/3 complex subunit 2 {ECO:0000250; ARC34; Arp2/3 complex 34 kDa subunit; arp2/3 complex 34 kDa subunit {ECO:0000250; ARP2/3 protein complex subunit 34; ARPC2; arpc2 {ECO:0000250; p34-ARC; p34-ARC {ECO:0000250; PNAS-139; PRO2446; testis tissue sperm-binding protein Li 53e; UniProtKB:O15144} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10109, 301511, 76709 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15144, P85970, Q9CVB6 | |
| ARPC2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ARPC2 (NP_6906011). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction