missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00
Specifications
| Antigen | ARPC1A |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18337216
|
Novus Biologicals
NBP3-04783-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18312997
|
Novus Biologicals
NBP3-04783-100UL |
100 μL |
N/A
|
N/A | |||||
Description
ARPC1A Polyclonal specifically detects ARPC1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ARPC1A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 10552 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| actin binding protein (Schizosaccharomyces pombe sop2-like), actin related protein 2/3 complex, subunit 1A (41 kD), actin related protein 2/3 complex, subunit 1A, 41kDa, actin-related protein 2/3 complex subunit 1A, SOP2Hs, SOP2LArc40, SOP2-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 171-350 of human ARPC1A (NP_006400.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title