missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC1A Polyclonal Antibody

ARPC1A Polyclonal Antibody
Brand: Invitrogen PA588022
This item is not returnable.
View return policy
Description
ARPC1A Polyclonal Antibody for Western Blot
Specifications
| ARPC1A | |
| Polyclonal | |
| Unconjugated | |
| ARPC1A | |
| 0610010H08Rik; 1110030K07Rik; 41kDa; AA407347; actin binding protein (Schizosaccharomyces pombe sop2-like); actin related protein 2/3 complex subunit 1A; actin related protein 2/3 complex subunit 1A, 41kDa; actin related protein 2/3 complex, subunit 1A; actin related protein 2/3 complex, subunit 1A, 41kDa; actin-related protein 2/3 complex subunit 1A; ARC1A; Arc40; Arpc1a; epididymis luminal protein 68; epididymis secretory protein Li 307; HEL-68; HEL-S-307; Sid 329; Sid32; Sid329; SOP2Hs; SOP2L; SOP2-like protein; suppressor of profilin/p41 of actin-related complex 2/3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10552 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.83 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92747 | |
| ARPC1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-370 of human ARPC1A (NP_0064002). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction