missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC1A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ARPC1A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ARPC1A Polyclonal antibody specifically detects ARPC1A in Human samples. It is validated for Western BlotSpecifications
| ARPC1A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 10552 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| actin binding protein (Schizosaccharomyces pombe sop2-like), actin related protein 2/3 complex, subunit 1A (41 kD), actin related protein 2/3 complex, subunit 1A, 41kDa, actin-related protein 2/3 complex subunit 1A, SOP2Hs, SOP2LArc40, SOP2-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-370 of human ARPC1A (NP_006400.2). MSLHQFLLEPITCHAWNRDRTQIALSPNNHEVHIYKKNGSQWVKAHELKEHNGHITGIDWAPKSDRIVTCGADRNAYVWSQKDGVWKPTLVILRINRAATFVKWSPLENKFAVGSGARLISVCYFESENDWWVSKHIKKPIRSTVLSLDWHPNNVLLAAGSCDFKCRVFSAYIKEVDEKPASTPWGSKMPFGQLMSEFGGSGTGGWVHGVSFSASGSRLAWVSHDSTVSVADASKSVQVSTLKTEFLPLLSVSFVSENSVVAAGHDCCPMLFNYDDRGCLTFVSKLDIPKQSIQRNMSAMERFRNMDKRATTEDRNTALETLHQNSITQVSIYEVDKQDCRKFCTTGIDGAMTIWDFKTLESSIQGLRIM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title