missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arp3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118867
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ACTR3.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
Specifications
| Arp3 | |
| Polyclonal | |
| Unconjugated | |
| ACTR3 | |
| 1200003A09Rik; Actin-like protein 3; actin-related protein 3; actin-related protein 3 homolog (yest); ACTR3; Arp3; ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3; ARP3 actin-related protein 3 homolog; ARP3 actin-related protein 3 homolog (yeast) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10096, 74117, 81732 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61158, Q4V7C7, Q99JY9 | |
| ACTR3 | |
| A synthesized peptide derived from human ACTR3(Accession P61158), corresponding to amino acid residues A376-S418. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction