missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arp3 Monoclonal Antibody (3G2C9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531604
This item is not returnable.
View return policy
Description
MA5-31604 has been tested in indirect ELISA.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
Specifications
| Arp3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P61158, Q99JY9 | |
| ACTR3 | |
| Purified recombinant fragment of human ACTR3 (AA: 287-418) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 3G2C9 | |
| Unconjugated | |
| ACTR3 | |
| 1200003A09Rik; Actin-like protein 3; actin-related protein 3; actin-related protein 3 homolog (yest); ACTR3; Arp3; ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3; ARP3 actin-related protein 3 homolog; ARP3 actin-related protein 3 homolog (yeast) | |
| Mouse | |
| Protein G | |
| RUO | |
| 10096, 74117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction