missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arp2 Recombinant Rabbit Monoclonal Antibody (23GB610), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554053
This item is not returnable.
View return policy
Description
ARP2 (actin-related protein 2) is an ATP-binding component of the ARP2/3 complex - a multiprotein complex that mediates actin polymerization upon stimulation by the nucleation-promoting factor (NPF). The ARP2/3 complex mediates the formation of branched actin networks in the cytoplasm and provides the force for cell motility. The ARP2/3 complex also promotes actin polymerization in the nucleus, thereby regulating gene transcription and repair of damaged DNA.
Specifications
| Arp2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ACTR2 | |
| 4921510D23Rik; AA409782; Actin-like protein 14D; Actin-like protein 2; Actin-related protein 14 Da; Actin-related protein 14D; Actin-related protein 2; Actr14D; Actr2; Arp14D; ARP2; ARP2 actin related protein 2 homolog; ARP2 actin-related protein 2; ARP2 actin-related protein 2 homolog; ARP2 actin-related protein 2 homolog (yeast); Arp2-PA; Arp2-PB; Arp2-PC; CG9901; CG9901-PA; CG9901-PB; CG9901-PC; D6Ertd746e; Dmel\CG9901; Dmel_CG9901; I79_003843; LD18955p | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10097, 289820, 66713 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB610 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61160, P61161, Q5M7U6 | |
| ACTR2 | |
| A synthetic peptide of human Arp2 (350-394AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction