missing translation for 'onlineSavingsMsg'
Learn More

ARP 100, Thermo Scientific Chemicals

Product Code. 15496229
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496229 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496229 Supplier Thermo Scientific Alfa Aesar Supplier No. J64151.#0

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

A selective inhibitor of MMP-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 704888-90-4
Color White
Molecular Formula C17H20N2O5S
MDL Number MFCD09971105
Quantity 5 mg
Synonym mmp-2 inhibitor iii, n-hydroxy-2-n-isopropoxybiphenyl-4-ylsulfonamido acetamide, n-hydroxy-2-n-isopropoxy-1,1'-biphenyl-4-ylsulfonamido acetamide, 2-isopropoxy-1,1′-biphenyl-4-ylsulfonyl-amino-n-hydroxyacetamide, n-2-biphenyl-4-ylsulfonyl-n-2-isopropyloxy-acetohydroxamic acid, n-hydroxy-2-n-isopropoxy 1,1'-biphenyl-4-sulfonamido acetamide, d0q4ta, 2-biphenyl-4-sulfonyl-isopropoxy-amino-n-hydroxy-acetamide, n-hydroxy-2-n-isopropoxy4-phenylbenzenesulfonamido acetamide, n-hydroxy-2-n-propan-2-yloxy 4-phenylbenzene sulfonamido acetamide
Solubility Information Soluble at 100mM in DMSO
InChI Key PHGLPDURIUEELR-UHFFFAOYSA-N
SMILES CC(C)ON(CC(=O)NO)S(=O)(=O)C1=CC=C(C=C1)C2=CC=CC=C2
IUPAC Name N-hydroxy-2-[(4-phenylphenyl)sulfonyl-propan-2-yloxyamino]acetamide
Molecular Weight (g/mol) 364.416
PubChem CID 10044321
Formula Weight 364.42
Physical Form Solid
Chemical Name or Material ARP 100
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.