missing translation for 'onlineSavingsMsg'
Learn More

Aromatic Hydrocarbons Mixture 898 200 ug/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30318329
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30318329

Brand: Dr. Ehrenstorfer DREGA09000898ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trimethylbenzene∣1,3,5-Trimethylbenzene∣4-Cymene∣Benzene∣Ethylbenzene∣Isopropylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣n-Butylbenzene∣o-Xylene (1,2-Dimethylbenzene)∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣sec-Butylbenzene∣Styrene∣tert-Butylbenzene∣Toluene
CAS 95-63-6, 108-67-8, 99-87-6, 71-43-2, 100-41-4, 98-82-8
Analyte or Component Names 1,2,4-Trimethylbenzene∣1,3,5-Trimethylbenzene∣4-Cymene∣Benzene∣Ethylbenzene∣Isopropylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣n-Butylbenzene∣o-Xylene (1,2-Dimethylbenzene)∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣sec-Butylbenzene∣Styrene∣tert-Butylbenzene∣Toluene
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4-Trimethylbenzene 200 μg/mL∣1,3,5-Trimethylbenzene 200 μg/mL∣4-Cymene 200 μg/mL∣Benzene 200 μg/mL∣Ethylbenzene 200 μg/mL∣Isopropylbenzene 200 μg/mL∣m-Xylene (1,3-Dimethylbenzene) 200 μg/mL∣Naphthalene 200 μg/mL∣n-Butylbenzene 200 μg/mL∣o-Xylene (1,2-Dimethylbenzene) 200 μg/mL∣Propylbenzene 200 μg/mL∣p-Xylene (1,4-Dimethylbenzene) 200 μg/mL∣sec-Butylbenzene 200 μg/mL∣Styrene 200 μg/mL∣tert-Butylbenzene 200 μg/mL∣Toluene 200 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.