missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arogenate Dehydratase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119806
This item is not returnable.
View return policy
Description
Positive Control: Rice tissue lysates, Arabidopsis thaliana tissue lysates. Subcellular Location: Chloroplast, Plastid.
Converts the prephenate produced from the shikimate-chorismate pathway into phenylalanine.
Specifications
| Arogenate Dehydratase | |
| Polyclonal | |
| Unconjugated | |
| ADT | |
| Synthetic peptide within Rice Arogenate dehydratase aa 100-150. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction