missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1221 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30320618
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320618 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320618 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010302ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.