missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1016, SPEX CertiPrep™

Product Code. 12968887 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12968887

Brand: SPEX Certiprep PCB1016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Aroclor 101612674-11-2200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 12674-11-2, 110-54-3
CAS Max % 99.98, 0.02
Packaging 2mL amber prescored glass ampule
Molecular Formula C12H7Cl3
Quantity 1 mL
Synonym aroclor 1016, 3,3',5-trichlorobiphenyl, 1,1'-biphenyl, 3,3',5-trichloro, pcb 36, 3,3',5-trichloro-1,1'-biphenyl, 3,3/',5-trichlorobiphenyl, arochlor 1016, chlorodiphenyl cl, ccris 8481, dsstox_cid_3883
InChI Key RIBGNAJQTOXRDK-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)C2=CC(=CC(=C2)Cl)Cl
IUPAC Name 1,3-dichloro-5-(3-chlorophenyl)benzene
Molecular Weight (g/mol) 257.538
PubChem CID 38037
Concentration or Composition (by Analyte or Components) 200μg/mL
Chemical Name or Material Aroclor 1016
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : n-hexane

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.