missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARNT2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92823-0.02ml
This item is not returnable.
View return policy
Description
ARNT2 Polyclonal antibody specifically detects ARNT2 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| ARNT2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ARNT protein 2, aryl-hydrocarbon receptor nuclear translocator 2, bHLHe1BHLHE1, Class E basic helix-loop-helix protein 1, KIAA0307aryl hydrocarbon receptor nuclear translocator 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 440-690 of human ARNT2 (NP_055677.3). QLQQQQAELEVHQRDGLSSYDLSQVPVPNLPAGVHEAGKSVEKADAIFSQERDPRFAEMFAGISASEKKMMSSASAAGTQQIYSQGSPFPSGHSGKAFSSSVVHVPGVNDIQSSSSTGQNMSQISRQLNQSQVAWTGSRPPFPGQQIPSQSSKTQSSPFGIGTSHTYPADPSSYSPLSSPATSSPSGNAYSSLANRTPGFAESGQSSGQFQGRPSEVWSQWQSQHHGQQSGEQHSHQQPGQTEVFQDMLPM | |
| 0.02 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 9915 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction