missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARMX2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15664835
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15664835 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15664835 Supplier Invitrogen™ Supplier No. PA550912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARMCX2 Polyclonal Antibody detects endogenous levels of ARMCX2 protein.

The armadillo (ARM) repeat family of proteins are related to the Drosophila melanogaster armadillo protein, a protein essential for wingless signal transduction. ARM proteins are involved in a variety of processes such as cell migration, cell proliferation, tissue maintenance and tumorigenesis, and they also function in signal transduction and the maintenance of overall cell structure. ARMCX2 (armadillo repeat containing, X-linked 2), also known as ALEX2, is a 632 amino acid single-pass membrane protein that contains three ARM repeats and is highly expressed in testis, brain, ovary, heart, colon, spleen and prostate, where it is thought to play a role in tumor suppression. The gene encoding ARMCX2 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1, 000 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARMCX2
Gene Accession No. 0
Gene Alias 3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2
Gene Symbols ARMCX2
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human ARMCX2 (aa 321-370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367903
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.