missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538812
This item is not returnable.
View return policy
Description
This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintece of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome. [provided by RefSeq, Jul 2008].
Specifications
| ARMX1 | |
| Polyclonal | |
| Unconjugated | |
| ARMCX1 | |
| ALEX1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 501619, 51309 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P291, Q5U310 | |
| ARMCX1 | |
| A synthetic peptide derived from the internal region of human ARMX1 | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction