missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARMX1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15790131 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790131 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790131 Supplier Invitrogen™ Supplier No. PA552183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DEESTDTSEI GVETVKGAKT NAGAGSGAKL QGDSEVKPEV SLGLEDCPGV KEKAHSGSHS GGGLEAKAKA LFNTLKEQAS AKAGKGARVG TISGNRTLAP SLPCP Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 57%.

This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMCX1
Gene Accession No. Q9P291
Gene Alias 3010033I09Rik; AD032; ALEX1; ARM protein lost in epithelial cancers on chromosome X 1; arm protein lost in epithelial cancers, X chromosome, 1; armadillo repeat containing, X-linked 1; armadillo repeat-containing X-linked protein 1; ARMCX1; GASP7; Protein ALEX1
Gene Symbols ARMCX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMX1. Recombinant protein control fragment (Product #RP-91893).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.