missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £460.00
Specifications
| Antigen | C10orf76 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407661
|
Novus Biologicals
NBP2-30398-25ul |
25ul |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18182214
|
Novus Biologicals
NBP2-30398 |
0.1 mL |
£460.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARMH3 Polyclonal specifically detects ARMH3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| C10orf76 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Chromosome 10 Open Reading Frame 76, UPF0668 Protein C10orf76 | |
| C10orf76 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q5T2E6 | |
| 79591 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LEGKLESLDGEELMKIKDNINCLFQHCIQALGEEHPIRVVNALQTLCALIRGVHQKNKSTSGFDIINMLMGFDKAELCMKNLM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts