missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARMET Polyclonal Antibody

Description
A suggested positive control is rat brain tissue lysate. PA5-20432 can be used with blocking peptide PEP-0549.
The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.
Specifications
Specifications
| Antigen | ARMET |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MANF |
| Gene Accession No. | P0C5H9, P55145, Q9CXI5 |
| Gene Alias | 3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET |
| Gene Symbols | MANF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?