missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARMET Polyclonal Antibody

Product Code. 12818612
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818612 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818612 Supplier Invitrogen™ Supplier No. PA520431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse brain tissue lysate. PA5-20431 can be used with blocking peptide PEP-0548.

The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMET
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MANF
Gene Accession No. P0C5H9, P55145, Q9CXI5
Gene Alias 3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET
Gene Symbols MANF
Host Species Rabbit
Immunogen a 12 amino acid peptide from near the carboxy terminus of human MANF.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315989, 74840, 7873
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.