missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARMET Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589198
This item is not returnable.
View return policy
Description
Immunogen sequence: SRALRPGDCE VCISYLGRFY QDLKDRDVTF SPATIENELI KFCREARGKE NRLCYYIGAT DDAATKIINE VSKPLAHHIP VEKICEKLKK KDSQICELKY DKQIDLSTVD LKKLRVKELK KILDDWGETC KGCAEKSDYI RKINELMPKY APKAASAR; Positive Samples: Jurkat, U251, MCF7.
The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.
Specifications
| ARMET | |
| Polyclonal | |
| Unconjugated | |
| MANF | |
| 3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 315989, 74840, 7873 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C5H9, P55145, Q9CXI5 | |
| MANF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-182 of human ARMET/ARP/MANF (NP_0060014). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction