missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMER Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77198-0.025mg
This item is not returnable.
View return policy
Description
ARMER Polyclonal specifically detects ARMER in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ARMER | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ADP-ribosylation factor-like 6 interacting protein, ADP-ribosylation factor-like 6 interacting protein 1, Aip-1, AIP1, apoptotic regulator in the membrane of the endoplasmic reticulum, ARL-6-interacting protein 1, ARL6IPaip-1, ARMER, KIAA0069ADP-ribosylation factor-like protein 6-interacting protein 1 | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Q15041 | |
| ARL6IP1 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the C-terminus of human ARMER. The immunogen is located within the last 50 amino acids of ARMER. Amino Acid Squence: KREINKLLKQKEKKNE | |
| 0.025 mg | |
| Apoptosis | |
| 23204 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction