missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMCX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25705-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ALEX family of proteins which may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and a single Armadillo (arm) repeat. Other proteins containing the arm repeat are involved in development, maintece of tissue integrity, and tumorigenesis. This gene is closely localized with other family members on the X chromosome. Three transcript variants encoding the same protein have been identified for this gene.Specifications
| ARMCX3 | |
| Polyclonal | |
| Unconjugated | |
| ARMCX3 | |
| ALEX3, ARMCX3, KIAA0443, Protein ALEX3, UNQ2517/PRO6007 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51566, 71703 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BHS6, Q9UH62 | |
| ARMCX3 | |
| ARMCX3 Fusion Protein Ag22501 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction