missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMCX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ARMCX1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ARMCX1 Polyclonal specifically detects ARMCX1 in Human samples. It is validated for Western Blot.Specifications
| ARMCX1 | |
| Polyclonal | |
| Rabbit | |
| Q9P291 | |
| 51309 | |
| Synthetic peptides corresponding to ARMCX1(armadillo repeat containing, X-linked 1) The peptide sequence was selected from the C terminal of ARMCX1. Peptide sequence DNIKNEGLASSRKEFSRSSLFFLFKESGVCVKKIKALANHNDLVVKVKVL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ALEX1armadillo repeat-containing X-linked protein 1, ARM protein lost in epithelial cancers on chromosome X 1, arm protein lost in epithelial cancers, X chromosome, 1, armadillo repeat containing, X-linked 1, DKFZp686P06199, Protein ALEX1 | |
| ARMCX1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title