Learn More
Invitrogen™ ARMC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590832
Description
Immunogen sequence: MSERCCSRYS SGASIGCTPT STQAKMVSKR IAQETFDAAV RENIEEFAMG PEEAVKEAVE QFESQGVDLS NIVKTAPKVS ADGSQEPTHD ILQMLSDLQE SVASSRPQEV SAYLTRFCDQ CKQDKACRFL AAQKGAYPII FTAWKLATAG DQGLLLQSLN ALSVLTDGQP DLLDAQGLQL LVATLTQNAD EADLTCSGIR CVRHACLKHE QNRQDLVKAG; Positive Samples: A-431, 293T, LO2, Mouse brain, Mouse liver, Mouse pancreas, Rat testis, Rat brain.
The function of this gene's protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.
Specifications
| ARMC6 | |
| Polyclonal | |
| Unconjugated | |
| Armc6 | |
| 2410153K17Rik; armadillo repeat containing 6; armadillo repeat-containing protein 6; Armc6; AW554412; R30923_1; RGD1306691 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 306352, 76813, 93436 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NXE6, Q8BNU0 | |
| Armc6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ARMC6 (NP_0011861251). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.